Learn More
Gestigkeit Harry Hotplate with Performance Control and Thermostatic Controller
Click to view available options
Dimensions (L x W x H):
358 x 358 x 170 mm
592 x 442 x 170 mm
Temperature (Metric):
130°C to 370°C
30°C to 110°C
50°C to 300°C
Weight (Metric):
13 kg
26 kg
Voltage:
230 V
400 V
Unit Size:
Each
Beskrivelse
- Electrical hotplates with variable temperature control, with or without a thermostatic controller. Robust construction for continuous perfomance. aluminum alloy hotplate.
- Because of the asymmetrical, long-term heating system (hotplate corners and edges are subjected to more heat) an even temperature is guaranteed over the entire heating surface.
- Asbest-free insulation. The housing is constructed from high-grade, stainless steel No. 1.4301, whereby the central section has been additionally sprayed in orange.
- Four adjustable feet ensure extra stability. Connecting cable approx. 1,7 m. long, 230 Volt/ 3200 Watt with Schuko plug.
Tekniske data
Tekniske data
| Body Material | Stainless Steel |
| Top Plate Material | Anodized Aluminum |
| Temperature (Metric) | 130°C to 370°C |
| Weight (Metric) | 26 kg |
| Wattage | 4000 W |
| Includes | Performance Control and Thermostatic Controller |
| Voltage | 400 V |
| Type | Hotplate |
| Dimensions (L x W x H) | 592 x 442 x 170 mm |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion