Learn More
IKA™ HOTPLATE C-MAG HP 10 with EU Plug
Product Code. 10446558 Shop All IKA Products
Change view
Click to view available options
Top Plate Material:
Ceramic
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Top Plate Material Unit Size
10446558 Ceramic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10446558 Supplier IKA™ Supplier No. 0003582000
  • ONLINE EXCLUSIVE PRICE
    11975.00 NOK
    11100.70 NOK / Pack of 1
    Save 874.30 NOK
    7% Off
View all promotions
In Stock
Hotplate made of ceramic glass which offers excellent chemical resistance.
  • Fixed safety circuit of 550°C
  • Hot Top indicator - hot surface warning to prevent burns
  • Exact temperature setting via digital display (LED)
  • Bushing according to DIN 12878 for connecting a contact thermometer, e.g. ETS-D5, enables precise temperature control
  • Digital error code display
  • Elevated control panel for protection against leaking liquids
TRUSTED_SUSTAINABILITY
Specifications
Top Plate Material Ceramic
Temperature (English) 122°F to 932°F
Size 10 x 10 in.
Temperature (Metric) 50°C to 500°C
Depth (Metric) 330 mm
Height (English) 4 in.
Length (English) 13.25 in.
Width (English) 11.75 in.
Height (Metric) 10.16 cm
Length (Metric) 33.655 cm
Weight (Metric) 5 kg
Width (Metric) 29.845 cm
Wattage 1500 W
Amperage 6.3 A
Electrical Requirements 230/120/100 V, 50/60 Hz, 1005 w
Frequency 50/60 Hz
Safety Features Fixed safety circuit of 550°C
Plug Type EU-Style
No. of Positions 1
Shape Square
Voltage 230 V
Type Hotplate
Warranty 2 Years
Display LED Digital