Description
Combine Thermo Scientific™ Hot Gas Bypass Temperature Controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.
Features:
- 120V or 220V
- Factory or field installed
Ordering Information:
Field installed option must be installed by a qualified professional.
Specifications
Specifications
| Description | Factory Installed |
| Type | Factory-Installed |
| For Use With (Equipment) | CryoPlus 1,2,3,4 |
| Voltage | 220 V |