missing translation for 'onlineSavingsMsg'
Learn More

HUBER Hose for Cooling Water

Braided hose

Brand:  HUBER 16852

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10555413

  • 977.32 NOK / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

  • Braided hose
  • Min. temp.: -20°C
  • Max. temp.: 90°C
Specifications

Specifications

Cooling Water PZ-90-1.5-G1/2
-20°C to +90°C
Hose
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers