Learn More
HUBER Hose for Cooling Water
Braided hose
Brand:  HUBER 16854
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 10339382
  • 977.32 NOK / Each
Estimated Shipment: 18-06-2024
Description
  • Braided hose
  • Min. temp.: -20°C
  • Max. temp.: 90°C
Specifications
Specifications
Cooling Water PZ-90-1-G3/4
-20°C to +90°C
Hose