Learn More
Reitenspiess™ Hose Cleaning Brush with Eyelet
Product Code. 16308685
Change view
Click to view available options
Bristle Material:
Natural
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bristle Material Unit Size
16308685 Natural Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16308685 Supplier Reitenspiess™ Supplier No. 31300501
  • ONLINE EXCLUSIVE PRICE
    158.79 NOK
    87.65 NOK / Pack of 1
    Save 71.14 NOK
    44% Off
Estimated Shipment: 11-01-2027
The Hose Cleaning Brush with Eyelet features a twisted galvanized wire handle with eyelet. Available with with nylon or natural bristles.
  • Twisted galvanized wire handle with eyelet
  • Choose between nylon or natural bristles
Specifica
Product Type Hose Brush
Bristle Material Natural
Handle Material Galvanized Wire
Handle Type Twisted Wire
Diameter (Metric) Bristle 30 mm
Length (Metric) 1000 mm
Length (Metric) Brush 150 mm
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.