Description
- Galvanised steel and aluminum
- For flexible hoses with outer Ø up to 10mm, 15mm, 20mm and 30mm
- Ensure that the stop-it metal is extremly heat resistant and sturdy
- With the hose clip stop-it's knurled screw, a stepless, delicate flow regulation or complete stop of the flow are possible, even for existing hose connections
- The hose is compressed over a wide area against the sturdy steel hook in order to prevent it from being damaged
Specifications
Specifications
| Type | Hose Clamp |
| Material | Galvanized Steel, Aluminum |
| For Use With (Equipment) | 15 mm O.D. Flexible Hoses |
| Description | Extremely heat resistant and sturdy |