Learn More
Buerkle™ Hose Angle Bracket with Strong Outer Thread
Shop All Bürkle Products
Click to view available options
Diameter (Metric):
12 mm nozzle
25 mm nozzle
Nozzle: 20 mm
Nozzle: 22 mm
Thread Size:
1 in.
1/2 in.
3/4 in.
Unit Size:
Each
Description
- Manufactured from polypropylene
- Fixed
- External thread
Specifications
Specifications
| Certifications/Compliance | EU Foodstuffs |
| Diameter (Metric) | Nozzle: 22 mm |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | Ball Valve |
| Thread Size | 3/4 in. |
| Width (Metric) | 17 mm |
| Product Type | Tube Bend |