Learn More
Hirschmann™ Hose Adapter
Description
- Spare part for pipetus™ standard, hose connectors to pump
Specifications
Specifications
| Color | Gray |
| For Use With (Equipment) | Pipetus Standard |