missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Hormones: 3,3',5-Triiodo-L-Thyronine
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1000 mg
Unit Size:
100mg
1g
250mg
500mg
Description
For research use only. Not intended for therapeutic use.
Specifications
Specifications
| Form | Crystalline powder |
| Product Type | Progesterone |
| Sterility | Sterile |
| pH | 9 |
| Quantity | 500 mg |
| Source | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction