Learn More
Scientific Industries SI™ Horizontal Tube Holder
Snap tubes into place swiftly and easily. Scientific Instruments SI™ Horizontal Tube Holder does not require additional attachments for use.
Brand: Scientific Industries SI™ SI-H524
Additional Details : Weight : 0.10000kg
Description
- Holds tubes in place horizontally.
- Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
- No additional attachments are necessary
- Holds tubes in place horizontally.
- Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
- No additional attachments are necessary
Specifications
24 Microtubes | |
Vortex-Genie 2 Mixer and the Vortex-Genie Pulse | |
24 |