Learn More
Scientific Industries SI™ Horizontal Tube Holder
Product Code. 10286369
Click to view available options
Holds:
12 x 15 mL Tubes
24 Microtubes
50 mL Tubes
Unit Size:
Each
Product Code. 10286369
Brand: Scientific Industries SI™ 83041276
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Snap tubes into place swiftly and easily. Scientific Instruments SI™ Horizontal Tube Holder does not require additional attachments for use.
  • Holds tubes in place horizontally.
  • Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
  • No additional attachments are necessary
  • Holds tubes in place horizontally.
  • Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
  • No additional attachments are necessary
Specifications
Holds 50 mL Tubes
For Use With (Equipment) Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
No. of Positions 6