Learn More
Scientific Industries Horizontal Orbital Shaker Attachment for Enviro-Genie and Incubator-Genie
Product Code. 16657762 Shop All Ohaus Products
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
16657762 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16657762 Supplier Scientific Industries Supplier No. 83041357
  • ONLINE EXCLUSIVE PRICE
    4467.75 NOK
    4080.76 NOK / Pack of 1
    Save 386.99 NOK
    8% Off
In Stock
An accessory that adds horizontal orbital shaking capability to the Enviro-Genie and the Incubator-Genie.
  • Adds horizontal orbital shaking capability to the Enviro-Genie and the Incubator-Genie with variable speed of 15-100 rpm
  • Ideal for Western Blots and for maintaining cells or solutions in suspension
  • No tools required
  • Simply remove the Magnetic Platform from the Enviro-Genie or Incubator-Genie and attach the Horizontal Orbital Shaker Attachment
  • Fits all new and existing Enviro and Incubator models
TRUSTED_SUSTAINABILITY
Specifications
Load Bearing Capacity (Metric) 2 kg
Holds Various containers including flasks and microplates
Depth (English) 13 in.
Depth (Metric) 330.20 mm
Dimensions (L x W) 13 x 13 in.
Dimensions (L x W x H) 13 x 13 x 5 in.
Height (English) 5 in.
Height (Metric) 127 mm
Length (English) 13 in.
Length (Metric) 330.20 mm
Width (English) 13 in.
Width (Metric) 330.20 mm
Weight (English) 3.5 lb.
Weight (Metric) 1.6 kg
Color Black
Description Horizontal Orbital Shaker for Enviro-Genie and Incubator-Genie
For Use With (Application) Orbital Shaking of Various Containers Including Flasks and Microplates.
For Use With (Equipment) Scientific Industries Enviro-Genie and Incubator-Genie
Includes Horizontal Orbital Shaker Attachment with Texture Mat
No. of Positions 1
Packaging Quantity 1 Ea.