Learn More
Fisherbrand™ Horizontal Large Gel Systems: Combs
Click to view available options
Teeth Count:
1
8
12
16
20
24
28
32
36
Thickness (Metric) Teeth:
0.1 cm
0.15 cm
20 cm
Unit Size:
Each
Description
For large gel systems
Specifications
Specifications
| For Use With (Equipment) | Owl A2 and A5 Large Gel Electrophoresis System |
| Type | Wall Comb |
| Teeth Count | 1 |
| Thickness (Metric) Teeth | 20 cm |
| Width (English) Teeth | 7.8 in. |
| Width (Metric) Teeth | 20 cm |