Learn More
BD Horizon™ Brilliant Stain Buffer
Description
Designed to complement multicolor flow cytometry experiments that utilize two or more different staining reagents conjugated with BD Horizon Brilliant fluorescent polymer dyes
Fluorescent dye interactions may cause staining artifacts which may affect data interpretation
For optimal and reproducible results, BD Horizon Brilliant Stain Buffer should be used anytime two or more BD Horizon Brilliant dyes are used in the same experiment
When using only one BD Horizon Brilliant reagent in an experiment, the Brilliant Stain Buffer is not needed
The BD Horizon Brilliant Stain Buffer is compatible with the use of other fluorescent staining reagents conjugated with traditional fluorochromes, such as fluorescein, phycoerythrin or Alexa Fluor™ dyes.
- Volume per test: 50μL/test
- 1000 tests
Specifications
Specifications
| Color | Colorless |
| Content And Storage | 4°C |
| For Use With (Application) | Immunofluorescence |
| Format | Liquid |
| Quantity | 1000 tests |