Description
- The Pocketless Hygiene gown protects the working environment from particles coming from the body of the operator
- Allows the user to be protected from the risks of soiling related to the activity being carried out
- Pleasant and light, polypropylene fabric is extremely breathable
- Collared model without flap and elasticated at the wrists
Specifications
Specifications
| Size | X-Large |
| Construction | Made in China |
| For Use With (Application) | Category 1 Personal Protective Equipment |
| Gender | Unisex |
| No. of Pockets | None |
| Packaging Type | Individual, Bag |
| Product Type | Hygiene Gown |
| Material | Polypropylene |
| Color | Red |
| Closure Type | Press Studs |
| Show More |