Learn More
Medicom™ Hopen™ Pocketless Hygiene Gown
Shop All Medicom Products
Click to view available options
Color:
Red
White
Size:
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
Description
- The Pocketless Hygiene gown protects the working environment from particles coming from the body of the operator
- Allows the user to be protected from the risks of soiling related to the activity being carried out
- Pleasant and light, polypropylene fabric is extremely breathable
- Collared model without flap and elasticated at the wrists
Spécification
Spécification
| Size | 3X-Large |
| Construction | Made in China |
| For Use With (Application) | Category 1 Personal Protective Equipment |
| Gender | Unisex |
| No. of Pockets | None |
| Packaging Type | Individual, Bag |
| Product Type | Hygiene Gown |
| Material | Polypropylene |
| Color | White |
| Closure Type | Press Studs |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu