Learn More
Medicom™ Hopen™ Pocketed Hygiene Gown
GREENER_CHOICE
Product Code. 16554061 Shop All Medicom Products
Change view
Click to view available options
Color:
White
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Size Unit Size
16554061 White 2X-Large Pack of 50
16524061 White Medium Pack of 50
16534061 White Large Pack of 50
16544061 White X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16554061 Supplier Medicom™ Supplier No. 40.401/05
  • ONLINE EXCLUSIVE PRICE
    666.00 NOK
    608.97 NOK / Pack of 50
    Save 57.03 NOK
    8% Off
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
Medicom™ Hopen™ Pocketed Hygiene Gown with press-stud fasteners, available in two colors
  • Gown protects the operator, as well as sensitive products/processes that could be contaminated in a protected atmosphere
  • Protects user against radioactive particles, and infectious agents
  • Reduces the risk of soiling related to the user's activity
  • Pleasant and light, polypropylene fabric is breathable yet durable
  • Variety of colors allows for possibility of identification by color codes
  • Two pockets: one on the chest, and one on the lower thigh
  • Individually packaged for controlled hygiene, and distribution
  • Pockets located on the upper left, and lower right of the gown
Specifications
Product Type Hygiene Gown
Size 2X-Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Press Studs
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Radioactive Particles, Infectious Agents
No. of Pockets 2
Seam Type Sewn
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
For Use With (Application) Category 1 Personal Protective Equipment
Packaging Type Individual, Bag
Construction Made in China