Learn More
Ohaus™ Hook, R21, RC21, R31, RC31, V71
Product Code. 15882932 Shop All Ohaus Products
Change view
Click to view available options
:
For use with: variety of compact and counting scales, 0.2 mL PCR Plate/Tube Thermal Block
Product Code.
15882932 For use with: variety of compact and counting scales, 0.2 mL PCR Plate/Tube Thermal Block
1 options
Product Code. 15882932
Brand: Ohaus™ 30037450
Counting/compact scale hook suitable for a variety of scale models.
  • Suitable for a variety of compact and counting scales.
    • Compact Scale, V71P1502T
    • Compact Scale, V71P3T
    • Compact Scale, V71P6T
    • Compact Scale, V71P15T
    • Compact Scale, V71P30T
    • Compact Scale, V71P1502T-M
    • Compact Scale, V71P3T-M
    • Compact Scale, V71P6T-M
    • Compact Scale, V71P15T-M
    • Compact Scale, V71P30T-M
    • Counting Scale, RC31P1502-M
    • Counting Scale, RC31P15-M
    • Counting Scale, RC31P30-M
    • Counting Scale, RC31P3-M
    • Counting Scale, RC31P6-M
    • Counting Scale, RC31P1502
    • Counting Scale, RC31P3
    • Counting Scale, RC31P6
    • Counting Scale, RC31P15
    • Counting Scale, RC31P30
    • Also fits 0.2 mL PCR Plate/Tube Thermal Block
TRUSTED_SUSTAINABILITY
Specifications
Product Type Metal Hook
Description Hook, R21 RC21 R31 RC31 V71
Classification Miscellaneous
For Use With (Equipment) Variety of compact and counting scales, 0.2 mL PCR Plate/Tube Thermal Block