Description
- Compact shippers have a cylindrical interior to hold tubes, vials and other small items
- Add as many ice packs as you need
- For non-hazardous materials only
Specifications
Specifications
| Height (English) | 6 in. |
| Length (English) | 6 in. |
| Width (English) | 6 in. |
| Dimensions (L x W x H) | 6 x 6 x 6in. |
| Diameter (English) | 3 in. |
| Type | Compact Insulated Shipper |
| Quantity | 1 Each |