Learn More
Sperian™ Honeywell™ HI-STYLE™ 6000 Clear Lens Safety Glasses
Description
- With anti-scratch coating
- Integral temple wire provides side arm adjustment
- General use and protection against impact (45m/s)
Specifications
Specifications
| Certifications/Compliance | EC 0035, EC 0086 |
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Eye and Face Protection Spectacles |
| For Use With (Application) | General use and protection against impact (45m/s) Mechanic work : Grinding, turning, milling |
| Frame Color | Cobalt Blue |
| No. per Pack | 1 Ea. |
| Lens Material | Polycarbonate |
| Frame Material | Polyamide |