Learn More
Sperian™ Honeywell™ Santa Cruz Safety Glasses
Description
- Adjustable positive-lock temples
- Polycarbonate lenses
- Provides 99.9% UV protection
- Available in blue or red
Specifications
Specifications
| Certifications/Compliance | ISO 9001 / 2000, EC EC0165 |
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Eye and Face Protection Spectacles |
| For Use With (Application) | UV protection glasses |
| Frame Color | Blue |
| No. per Pack | 1 Ea. |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |