Learn More
Sperian™ Honeywell™ A700 Series Clear Anti-Fog Safety Glasses
Description
- Minimizes lens fogging in extreme conditions
- Affordably priced and economical for your business
- Lightweight design with sport temples and soft temple tip pads
- Moulded universal nose bridge
- Provides a secure, wraparound fit for all-day comfort
- 8-base polycarbonate abrasion-resistant lenses
- Provides a 180° field of distortion-free vision
Specifications
Specifications
| Lens Coating | Anti-fog |
| Product Type | Safety Glasses |
| Frame Color | Clear |
| No. per Pack | 10/Pk. |
| Lens Material | Polycarbonate |