Learn More
Sperian™ Honeywell™ 2000 Comfort Series Respirators
Product Code. 15341141
Change view
Click to view available options
Model:
2110 FFP1
2110 FFP2
2211 FFP2V
Unit Size:
Pack of 20
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
15341141 2110 FFP1 Pack of 25
15371141 2211 FFP2V Pack of 20
15361141 2110 FFP2 Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15341141 Supplier Sperian™ Supplier No. 1031590
  • ONLINE EXCLUSIVE PRICE
    354.49 NOK
    288.50 NOK / Pack of 25
    Save 65.99 NOK
    18% Off
Estimated Shipment: 15-12-2026
With an ergonomic design to deliver outstanding levels of comfort and adjustment. Sperian™ Honeywell™ 2000 Comfort Series Respirators have a cut-out shape to provide excellent compatibility with goggles.
  • Folds flat packed in individually sealed bags for transportation and storage
  • Dolomite tested, demonstrating the respirators' resistance to clogging over time
  • With high efficiency exhalation valve for protection
  • Double layer face seal provides comfort and protection
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 149 : 2001 A1: 2009
Disposable Single-use
Filter Type FFP1
Product Type Respirator Protection
Size Medium
With Exhalation Valve Yes
For Use With (Application) Agriculture, Chemical, Construction, Fire Protection, Food Services, Forestry, Government, Manufacturing, Mining, Municipal Services, Oil and Gas, Pharmaceutical, Steel and Metals, Utilities
Latex Free Yes
Model 2110 FFP1
Quantity 25/Pk.
Shape Flat Fold