missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homosalate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Homosalate-d4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 D4 H18 O3 |
| Quantity | 1 mg |
| Synonym | Salicylic Acid 3,3,5-Trimethylcyclohexyl Ester-d4, 3,3,5-Trimethylcyclohexanol Salicylate-d4, 3,3,5-Trimethylcyclohexyl Salicylate-d4, Eusolex HMS-d4, Filtrosol A-d4, Heliopan-d4, Heliopan HMS-d4, Heliophan-d4, Homomenthyl Salicylate-d4, Kemester-d4, Kemester HMS-d4, NSC 164918-d4, Neo Heliopan-d4, Neo Heliopan HMS-d4, Parsol HMS-d4, Uniderm Homsal-d4 |
| InChI Formula | InChI=1S/C16H22O3/c1-11-8-12(10-16(2,3)9-11)19-15(18)13-6-4-5-7-14(13)17/h4-7,11-12,17H,8-10H2,1-3H3/i4D,5D,6D,7D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OC2CC(C)CC(C)(C)C2)c(O)c1[2H] |
| IUPAC Name | (3,3,5-trimethylcyclohexyl) 2,3,4,5-tetradeuterio-6-hydroxy-benzoate |
| Molecular Weight (g/mol) | 266.37 |
| Formula Weight | 266.182 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?