Learn More
Alfa Aesar™ Holmium, plasma standard solution, Specpure™, Ho 1000μg/mL
Brand: Alfa Aesar™ 035756.AD
Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Quantity | 50mL |
---|
Chemical Identifiers
MFCD00011049 |
Specifications
Holmium plasma standard solution | |
MFCD00011049 | |
Ho2O3 in 5% HNO3 | |
Specpure™ |
Ho 1000μg/mL | |
UN3264 | |
Liquid | |
50mL |