Learn More
Holmium(III) 2,4-pentanedionate, REacton™, 99.9% (REO)
Product Code. 11342298
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. 11342298
Brand: Thermo Scientific Alfa Aesar 013199.09
Request Bulk or Custom Format
CAS: 14589-33-4 | C15H21HoO6 | 462.26 g/mol
It is used for Building Blocks of chemicals and , Chemical Synthesis, used as a trihydrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14589-33-4
Molecular Formula C15H21HoO6
Molecular Weight (g/mol) 462.26
MDL Number MFCD00050176
InChI Key UEKRGRZSLATUQV-UHFFFAOYSA-N
Synonym holmium 2,4-pentanedionate, holmium iii 2,4-pentanedionate, reacton, holmium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy holmio oxy pent-3-en-2-one
PubChem CID 24282829
IUPAC Name holmium;(Z)-4-hydroxypent-3-en-2-one
SMILES [Ho+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Specifications
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 462.26
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Holmium(III) 2,4-pentanedionate