Learn More
DWK Life Sciences Holding Device for Reaction Vessels, from chrome-nickel steel
Tienda DWK Life Sciences ProductosDescripción
Reaction vessels can be operated and tempered in various ways. To prevent the reaction apparatus from moving during the test, the reaction vessel should ideally be suspended from a tripod linkage by means of a holding device. Cooling and heating media can thus be fed flexibly from below to the reaction vessel and removed again.
- For fixed suspension of the lid and the reaction vessel on a tripod rod
Especificaciones
Especificaciones
| Description | DN 150 |
| Material | Chrome-Nickel Steel |
| For Use With (Application) | Holder for lids and reaction vessels; Mounting |
| No. per Pack | 1/Pk. |
| Autoclavable | Not Autoclavable |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.