Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Sponges
Description
- 3 or 6 mm Thick
- 4 pcs
Specifications
Specifications
| For Use With (Equipment) | TE 42, TE 62 Transfer Tank |
| Type | Sponge |
| Material | Dacron |
| Quantity | 4 |
| Thickness (English) | 0.25 in. |
| Thickness (Metric) | 0.6 cm |