Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Electrode Panels
Shop All Cytiva ProductsDescription
- Comes in gray or black electrode panels
- Type: Gray Electrode Panel
- Includes Banana plug and platinum wire
Specifications
Specifications
| Color | Gray |
| For Use With (Equipment) | TE 42, TE 62 Transfer Tank |
| Type | Gray Electrode Panel |
| Includes | Banana plug and Platinum wire |