Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Electrode Panels
Product Code. 12054747 Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
TE 22 Mini Tank Transfer Unit
TE 42, TE 62 Transfer Tank
Type:
Black Electrode Panel
Electrode Panel
Gray Electrode Panel
Unit Size:
Each
Product Code. 12054747
Brand: Hoefer™ TE43BK
Only null left
Add to basket
Add to basket
For use with TE 42 and TE 62 Transfer Tank
  • Comes in gray or black electrode panels
  • Type: Gray Electrode Panel
  • Includes Banana plug and platinum wire
Specifications
Color Black
For Use With (Equipment) TE 42, TE 62 Transfer Tank
Type Black Electrode Panel
Includes Banana plug and Platinum wire