Learn More
Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates
Product Code. 10380735
Change view
Click to view available options
Type:
Low Fluorescence Glass Plate
Low Fluorescence Notched Glass Plate
Notched Alumina Plate
Material:
Alumina
Glass
Quantity:
1/Pk.
10/Pk.
5/Pk.
Unit Size:
Each
Pack of 10
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Material Quantity Unit Size
10380735 Low Fluorescence Glass Plate Glass 10/Pk. Pack of 10
10473162 Low Fluorescence Notched Glass Plate Glass 5/Pk. Pack of 5
12034727 Notched Alumina Plate Alumina 1/Pk. Each
10585585 Notched Alumina Plate Alumina 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10380735 Supplier Hoefer™ Supplier No. SE202P10
  • ONLINE EXCLUSIVE PRICE
    1020.00 NOK
    813.66 NOK / Pack of 10
    Save 206.34 NOK
    20% Off
Estimated Shipment: 02-12-2026
Use with the SE 250 vertical electrophoresis unit. Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates come in a variety of styles for particular needs.
  • 10 x 8 cm
Specifications
For Use With (Equipment) SE 250 Vertical Electrophoresis Unit
Type Low Fluorescence Glass Plate
Material Glass
Quantity 10/Pk.
Shape Rectangular