Learn More
Hoefer™ Easy Breeze™ Gel Dryer Accessories: Film
Click to view available options
For Use With (Equipment):
Standard Frame of the SE 1200 Easy Breeze Air Gel Drying System
Unit Size:
Pack of 50
Description
Gel Drying Film
- For photography or fluorography, clear cellophane ensures a photograph or exposure without distortion
- For autoradiography, use of a Mylar™ sheet in place of one cellophane sheet allows the Mylar to be easily removed after drying for maximum beta emission
Specifications
Specifications
| For Use With (Equipment) | Standard Frame of the SE 1200 Easy Breeze Air Gel Drying System |
| Type | Porous Cellophane Sheet |