Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Product Code. 12004657
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Reference
10
15
18
5
9
Thickness (Metric):
0.07 cm
0.075 cm
0.1 cm
0.15 cm
Volume (Metric) Well:
120μL/ mm
13μL/ mm
19.5μL/ mm
2.2μL/ mm
2.9μL/ mm
4.4μL/ mm
4.8μL/ mm
5.8μL/ mm
51μL/ mm
68μL/ mm
7.2μL/ mm
9.8μL/ mm
Unit Size:
Each
Product Code. No. of Wells Thickness (Metric) Volume (Metric) Well unitSize
12004657 1 Prep, 1 Reference 0.15 cm 120μL/ mm Each
12019847 9 0.1 cm 5.8μL/ mm Each
10289444 18 0.1 cm 2.9μL/ mm Each
10041742 18 0.1 cm 2.9μL/ mm Each
12904830 15 0.07 cm 2.2μL/ mm Each
11549663 15 0.07 cm 2.2μL/ mm Each
12059807 1 Prep, 1 Reference 0.07 cm 51μL/ mm Each
12004797 5 0.07 cm 9.8μL/ mm Each
12084847 10 0.075 cm 9.8μL/ mm Each
11569663 15 0.15 cm 4.4μL/ mm Each
12079857 15 0.1 cm 2.9μL/ mm Each
12074697 10 0.15 cm 7.2μL/ mm Each
10209834 10 0.15 cm 7.2μL/ mm Each
12054797 5 0.15 cm 19.5μL/ mm Each
12094797 10 0.1 cm 4.8μL/ mm Each
12044797 5 0.1 cm 13μL/ mm Each
12044737 1 Prep, 1 Reference 0.1 cm 68μL/ mm Each
17 options
Product Code. 12004657
Brand: Hoefer™ SE211AR1.5
Only null left
Add to basket
Add to basket
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
  • Lightweight
  • Virtually unbreakable
Specifications
For Use With (Equipment) SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
Type Comb
Material PTFE
No. of Wells 1 Prep, 1 Reference
Thickness (English) 0.05 in.
Thickness (Metric) 0.15 cm
Volume (Metric) Well 120μL/ mm
Width (English) Well 2.6 in.
Width (Metric) Well 6.8 cm
Depth (English) 0.51 in.
Depth (Metric) 1.3 cm