Learn More
Hoefer™ Blotting Paper
Product Code. 12014757 Shop All Cytiva Products
Click to view available options
Length (Metric):
9 cm
14 cm
14.5 cm
21 cm
Width (Metric):
10.5 cm
16 cm
21.5 cm
25 cm
For Use With (Equipment):
TE22 Mini Tank or MiniVE Module
TE42 or TE62 Tank Blotting Units
TE70 or TE77 Semi-Dry Blotting Unit
TE77 Semi-Dry Blotting Unit
Unit Size:
Pack of 25
Pack of 50
Product Code. 12014757
Brand: Hoefer™ TE46
Only null left
Add to basket
Add to basket
Cellulose papers precut for convenience
  • For Western Blotting
Specifications
Length (Metric) 14.5 cm
Width (Metric) 21.5 cm
For Use With (Equipment) TE42 or TE62 Tank Blotting Units
Format Sheet
Material Cellulose