Learn More
MedChemExpress Hoechst 33342 analog 2 trihydrochloride
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 20 mg/mL (30.31 mM; Need ultrasonic) ∣ DMSO : 4 mg/mL (6.06 mM; Need ultrasonic) |
| Molecular Weight (g/mol) | 659.78 g/mol |
| CAS | 155815-98-8 |