Learn More
Karl Hecht Shoulder bottles made of polyethylene 250ml
Brand:  Karl Hecht 49105024
Shoulder bottles made of polyethylene 250ml
Product Code. 10014141
  • ONLINE EXCLUSIVE PRICE
    26.25 NOK
    24.32 NOK / Each
    Save 1.93 NOK
    7% Off
Only null left
Estimated Shipment: 27-01-2026