missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ hnRNP K Antibody (F45 P9 C7), Novus Biologicals™

Product Code. 30066982 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
50 μg
Unit Size:
200µg
50µg
Product Code. Quantity unitSize
30066982 200 μg 200µg
30067084 50 μg 50µg
2 options
This item is not returnable. View return policy

Product Code. 30066982

Brand: Novus Biologicals™ NBP330893200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

hnRNP K Monoclonal antibody specifically detects hnRNP K in Chicken,Human,Mouse,Rabbit,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP K
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone F45 P9 C7
Conjugate Unconjugated
Dilution Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-5 μg/mL
Formulation Phosphate buffered saline
Gene Alias dC-stretch binding protein, FLJ41122, heterogeneous nuclear ribonucleoprotein K, hnRNP K, transformation upregulated nuclear protein, Transformation up-regulated nuclear protein, TUNPHNRPKCSBP
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the C-terminal of the hnRNP K protein and mouse myeloma Ag8563 cells. Sequence common in human, mouse, rat, rabbit and chicken
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3190
Target Species Chicken, Human, Mouse, Rabbit, Rat
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.