missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hnRNP F Monoclonal Antibody (3H4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA134551
This item is not returnable.
View return policy
Description
MA1-34551 detects HNRNP F from human samples.
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins. The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA. These proteins are associated with pre-mRNAs in the nucleus and regulate alternative splicing, polyadenylation, and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs which have guanosine-rich sequences. This protein is very similar to the family member hnRPH. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| hnRNP F | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P52597 | |
| Hnrnpf | |
| Full length hnRNP F protein (human). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 3H4 | |
| Unconjugated | |
| Hnrnpf | |
| 4833420I20Rik; AA407306; heterogeneous nuclear ribonucleoprotein F; Heterogeneous nuclear ribonucleoprotein F, N-terminally processed; hnRNP F; HnRNP F protein; hnRNP H; Hnrnpf; HNRNPH1; Hnrpf; mcs94 1; mcs94-1; Nucleolin-like protein mcs94-1; OK/SW-cl.23; ribonucleoprotein F; ROF | |
| Mouse | |
| Protein A | |
| RUO | |
| 3185 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction