missing translation for 'onlineSavingsMsg'
Learn More

HNF4G Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. p-7223525 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16844355 20 μL 20µL
16834355 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16844355

Brand: Proteintech 258011AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The HNF4G antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human HNF4G. This antibody recognizes human, mouse antigen. The HNF4G antibody has been validated for the following applications: FC, WB, ELISA analysis.

Specifications

Antigen HNF4G
Applications ChIP Assay, Flow Cytometry, Immunohistochemistry
Classification Polyclonal
Concentration 373 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 3174
Target Species Human, Mouse
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.