Learn More
HNF1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12533-1-AP-150UL
 View more versions of this product
Product Code. 16860284
  • 4836.10 NOK / 150µL
Description
This is a rabbit polyclonal antibody raised against the N-terminal region of human HNF1B. HNF1B exists some isoforms with MW of 61 kDa, 58 kDa, 46 kDa, and 44 kDa.
Hepatocyte Nuclear Factor 1 (HNF1, Homeoprotein LFB3, Transcription factor 2, TCF2, Variant hepatic nuclear factor) is a member of the homeodomain-containing superfamily of transcription factors. It is a liver-specific factor of the homeobox-containing basic helix-turn-helix family. The protein binds to DNA as either a homodimer, or a heterodimer with the related protein hepatocyte nuclear factor 1-alpha. HNF1has been shown to function in nephron development and regulates development of the embryonic pancreas. Mutations in the gene result in renal cysts and diabetes syndrome and noninsulin-dependent diabetes mellitus, and expression of the gene is altered in some types of cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
HNF1B
Polyclonal
Unconjugated
HNF1B
FJHN, HNF 1 beta, HNF 1B, HNF1 homeobox B, HNF1B, HNF1beta, HNF2, Homeoprotein LFB3, HPC11, LF B3, LFB3, MODY5, TCF 2, TCF2, Transcription factor 2, VHNF1
Rabbit
Antigen Affinity Chromatography
RUO
21410, 25640, 6928
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P23899, P27889, P35680
Hnf1b
HNF1B Fusion Protein Ag3232
150 μL
Primary
Human, Mouse, Rat