Learn More
Abnova™ HNF1B/CEN17q FISH Probe
Product Code. 16063375
Change view
Click to view available options
Quantity:
100 uL
200 uL
Unit Size:
100µL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16063375 100 uL 100µL
16073375 200 uL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16063375 Supplier Abnova™ Supplier No. FG0230.100uL
  • ONLINE EXCLUSIVE PRICE
    20550.00 NOK
    19713.21 NOK / 100µL
    Save 836.79 NOK
    4% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Specifications
Quantity 100 uL
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
For Use With (Application) FISH-Ce
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe