Learn More
HMHA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14832-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
HMHA1 (ARHGAP45) Contains a GTPase activator for the Rho-type GTPases (RhoGAP) domain that would be able to negatively regulate the actin cytoskeleton as well as cell spreading. However, also contains N-terminally a BAR-domin which is able to play an autoinhibitory effect on this RhoGAP activity.Specifications
HMHA1 | |
Polyclonal | |
Unconjugated | |
ARHGAP45 | |
HA 1, HLA HA1, HMHA1, KIAA0223, mHag HA 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23526, 314618, 70719 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3TBD2, Q92619 | |
Arhgap45 | |
HMHA1 Fusion Protein Ag6636 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |