Learn More
HLA-DR Mouse, anti-Human, PE, Clone: LN3, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60164PE
Encompass_Preferred
 View more versions of this product
Product Code. 17109231
  • 5476.63 NOK / 100 tests
Description
  • The LN3 antibody reacts with the HLA-DR antigen, a major histocompatibility complex (MHC) class II antigen encoded within the human leukocyte antigen (HLA) complex on chromosome 6
  • HLA-DR is a heterodimeric transmembrane glycoprotein composed of one α (36 kDa) and one β (27 kDa) subunit
  • MHC class II plays a central role in the presentation of antigen-derived peptides to CD4+ T cells, along with CD3/TCR and CD4
  • HLA-DR is primarily expressed on the surface of antigen presenting cells, including B cells, dendritic cells, monocytes, macrophages, thymic epithelial cells, and activated T cells
Specifications
Specifications
HLA-DR
Monoclonal
PE
100 Tests
Rhesus Monkey, Human
B Cells, Dendritic Cells, Macrophages, Monocytes, Other, T Cells
Immunohistochemistry, Flow Cytometry, Functional Assay
LN3
Mouse
Primary
IgG2b κ
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.