Learn More
HLA-DR Mouse, anti-Human, PE, Clone: LN3, STEMCELL Technologies
Product Code. 17119231
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
100 tests
25 tests
Product Code. Quantity unitSize
17119231 25 Tests 25 tests
17109231 100 Tests 100 tests
2 options
Product Code. 17119231
Brand: Stemcell Technologies 60164PE.1
Mouse Monoclonal Antibody
  • The LN3 antibody reacts with the HLA-DR antigen, a major histocompatibility complex (MHC) class II antigen encoded within the human leukocyte antigen (HLA) complex on chromosome 6
  • HLA-DR is a heterodimeric transmembrane glycoprotein composed of one α (36 kDa) and one β (27 kDa) subunit
  • MHC class II plays a central role in the presentation of antigen-derived peptides to CD4+ T cells, along with CD3/TCR and CD4
  • HLA-DR is primarily expressed on the surface of antigen presenting cells, including B cells, dendritic cells, monocytes, macrophages, thymic epithelial cells, and activated T cells
Specifications
Antigen HLA-DR
Applications Immunohistochemistry, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone LN3
Conjugate PE
Host Species Mouse
Quantity 25 Tests
Primary or Secondary Primary
Target Species Rhesus Monkey, Human
Isotype IgG2b κ
Cell Type B Cells, Dendritic Cells, Macrophages, Monocytes, Other, T Cells