missing translation for 'onlineSavingsMsg'
Learn More

HLA-DQA2 Monoclonal Antibody (OTI3E6), TrueMAB™, OriGene

Product Code. 17537534
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17537534 100 μL 100µL
17557534 30 μL 30µL
2 options
This item is not returnable. View return policy

Product Code. 17537534

Brand: Origene Technologies TA812935

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HLA-DQA2 Monoclonal antibody specifically detects HLA-DQA2 in Human samples. It is validated for Western Blot

Specifications

Antigen HLA-DQA2
Applications Western Blot
Classification Monoclonal
Clone OTI3E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HLA-DQA2
Gene Accession No. P01906
Gene Alias DQ alpha 2; DX ALPHA; DX alpha chain; DX-ALPHA; HLA class II histocompatibility antigen, DQ alpha 2 chain; HLA class II histocompatibility antigen, DQ(6) alpha chain; HLA DXA; HLA-DQA1; HLA-DQA2; HLA-DXA; major histocompatibility complex, class II, DQ alpha 2; MHC class II DQA2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 24-125 of human HLA-DQA2 (NP_064440) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3118
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.