missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA ABC Antibody (W6/32) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-64775JF635
This item is not returnable.
View return policy
Description
HLA ABC Monoclonal antibody specifically detects HLA ABC in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin) (Negative),CyTOF,Immunocytochemistry/ Immunofluorescence
Specifications
| HLA ABC | |
| Monoclonal | |
| Janelia Fluor 635 | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A&1, MHC class I antigen A&11, MHC class I antigen A&80 | |
| Purified human tonsil lymphocyte membranes | |
| 0.25 mL | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) (Negative), CyTOF, Immunocytochemistry/Immunofluorescence | |
| W6/32 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction