Learn More
Cytiva HiTrap™ IgG Sepharose FF
Product Code. 15539244 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15539244 5 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15539244 Supplier Cytiva Supplier No. 28908366
 Restricted Item
Estimated Shipment: 16-12-2026
Cytiva HiTrap™ IgG Sepharose FF is easy to use with a syringe, peristaltic pump, or a chromatography system.
  • Manual or system purification
  • Small-column cartridge
  • Supplied in a pack of 5 x 5 mL
TRUSTED_SUSTAINABILITY
Specifications
Bed Volume 5 mL
Diameter (Metric) Inner 16 mm
Quantity 5
Height (Metric) 25 mm
Storage Requirements 4 to 30°C, 20% Ethanol