Learn More
Cytiva HiTrap™ Capto™ MMC Impres Multimodal Chromatography Column
Product Code. 15181546 Shop All Cytiva Products
Change view
Click to view available options
Particle Size:
40 μm
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size Unit Size
15181546 40 μm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15181546 Supplier Cytiva Supplier No. 17371610
 Restricted Item
Estimated Shipment: 18-11-2026
Ready-to-use column for screening and small-scale protein purifications using ion exchange chromatography (IEX)
  • High-resolution beads and ligand selectivity ensures high yields
  • Enables use of platform approach to MAb process development
  • Displays broad pH/salt operational window
  • For screening of selectivity, binding, and elution conditions
  • Removes contaminants such as DNA, host cell proteins (HCP), leached protein A, and aggregates
  • Can be used in initial polishing step in MAb purification processes, as well as for polishing of antibody fragments such as domain antibodies
Avoid using oxidizing agents, cationic detergents.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) HPLC, MAbs Purification
Max. Pressure 300,000 psi
Particle Size 40 μm
compliance-icons
Product Identifier
  • hitrap capto MMC impres
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol