Learn More
Cytiva HiTrap Capto Butyl ImpRes
Product Code. 15497830 Shop All Cytiva Products
Change view
Click to view available options
Bed Volume:
1 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Volume Unit Size
15497830 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
The temperature difference between the fluid running through the column and the ambient temperature in the room should never be greater than 20°C
Product Code. 15497830 Supplier Cytiva Supplier No. 17371911
 Restricted Item
Estimated Shipment: 13-11-2026
The temperature difference between the fluid running through the column and the ambient temperature in the room should never be greater than 20°C
A ready-to-use column prepacked with BioProcess Capto Butyl ImpRes resins for screening and small-scale protein purification using hydrophobic interaction chromatography.
  • Prepacked columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
  • High-throughput chromatography resin for intermediate and polishing steps in a downstream protein purification process.
  • Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide.
  • High binding capacity, high flow rate, and low back pressure reduces process cycle times and improves productivity at process scale.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Bed Volume 1 mL
    Flow Rate Up to 220 cm/h in a 1 m diameter column with a bed height of 20 cm at 20°C; measured using process buffers with the same viscosity as water at 300 kPa.
    For Use With (Application) Hydrophobic Interaction Chromatography 
    Height (Metric) 25 mm
    Ligand Type Butyl
    Max. Pressure 5 bar [0.5 MPa] (70 psi)
    Particle Size 40 μm
    pH Range pH 3 to 13
    Storage Requirements 20% ethanol at 4°C to 30°C
    compliance-icons
    Product Identifier
    • HiTrap Capto Butyl ImpRes 1 ml, 5 x 1 ml
    Signal Word
    • Warning
    Hazard Category
    • Flammable liquid Category 3
    Hazard Statement
    • H226-Flammable liquid and vapour.
    Precautionary Statement
    • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
    Supplemental information
    • MIXTURE LIST-Contain: ethanol